Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057449.4 | NP_476797.1 | 1 - 2 | 233 | 96 | |
GenBank mRNA | AY061450.1 | - | 1 - 2 | 184 | 96 | |
X57485.1 | - | 193 | 96 | |||
X57496.1 | - | 203 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |