Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: retn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299844.1 | NP_001286773.1 | 2 - 3 | 826 | 79 | |
NM_057516.4 | NP_476864.2 | 2 - 3 | 826 | 79 | ||
NM_176254.2 | NP_788434.1 | 2 - 3 | 826 | 79 | ||
GenBank mRNA | AY118955.1 | - | 2 - 3 | 795 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |