Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057518.4 | NP_476866.1 | 1 - 2 | 236 | 97 | |
GenBank mRNA | BT132894.1 | - | 1 - 2 | 158 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |