Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pigeon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201915.2 | NP_001188844.1 | 5 - 6 | 2254 | 92 | |
NM_057598.5 | NP_476946.1 | 5 - 6 | 2254 | 92 | ||
GenBank mRNA | AY061465.1 | - | 5 - 6 | 2243 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |