Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299267.1 | NP_001286196.1 | 4 - 5 | 771 | 65 | |
NM_057769.4 | NP_477117.2 | 3 - 4 | 674 | 65 | ||
GenBank mRNA | AY058518.1 | - | 3 - 4 | 650 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |