Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057884.5 | NP_477232.1 | 6 - 7 | 1595 | 76 | |
GenBank mRNA | AF043263.1 | - | 6 - 7 | 1557 | 76 | |
BT044099.1 | - | 6 - 7 | 1594 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |