Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169159.2 | NP_001162630.1 | 10 - 11 | 6363 | 96 | |
NM_001169160.2 | NP_001162631.1 | 11 - 12 | 5731 | 96 | ||
NM_001297827.1 | NP_001284756.1 | 12 - 13 | 5988 | 96 | ||
NM_001297828.1 | NP_001284757.1 | 11 - 12 | 5853 | 96 | ||
NM_001297829.1 | NP_001284758.1 | 10 - 11 | 5697 | 96 | ||
NM_057941.5 | NP_477289.5 | 11 - 12 | 5800 | 96 | ||
NM_057942.5 | NP_477290.6 | 11 - 12 | 5917 | 96 | ||
NM_134314.4 | NP_599141.5 | 11 - 12 | 5865 | 96 | ||
NM_134315.4 | NP_599142.5 | 11 - 12 | 5917 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |