Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169159.2 | NP_001162630.1 | 13 - 14 | 7294 | 80 | |
NM_001169160.2 | NP_001162631.1 | 14 - 15 | 6662 | 80 | ||
NM_001297827.1 | NP_001284756.1 | 15 - 16 | 6919 | 80 | ||
NM_001297828.1 | NP_001284757.1 | 14 - 15 | 6784 | 80 | ||
NM_001297829.1 | NP_001284758.1 | 13 - 14 | 6628 | 80 | ||
NM_057941.5 | NP_477289.5 | 14 - 15 | 6731 | 80 | ||
NM_057942.5 | NP_477290.6 | 15 - 16 | 6872 | 80 | ||
NM_134314.4 | NP_599141.5 | 14 - 15 | 6796 | 80 | ||
NM_134315.4 | NP_599142.5 | 14 - 15 | 6848 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |