Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tutl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169397.2 | NP_001162868.1 | 7 - 8 | 1802 | 99 | |
NM_001169398.1 | NP_001162869.1 | 7 - 8 | 1750 | 99 | ||
NM_001273090.1 | NP_001260019.1 | 8 - 9 | 1935 | 99 | ||
NM_001273091.1 | NP_001260020.1 | 7 - 8 | 1802 | 99 | ||
NM_001316378.1 | NP_001303307.1 | 7 - 8 | 1802 | 99 | ||
NM_001316379.1 | NP_001303308.1 | 7 - 8 | 1802 | 99 | ||
NM_080127.6 | NP_524866.4 | 7 - 8 | 1802 | 99 | ||
NM_164576.4 | NP_722967.2 | 7 - 8 | 1802 | 99 | ||
NM_164577.3 | NP_722968.1 | 7 - 8 | 1802 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |