Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258575.1 | NP_001245504.1 | 1 - 2 | 630 | 90 | |
NM_080315.3 | NP_525054.2 | 1 - 2 | 630 | 90 | ||
GenBank mRNA | BT060443.1 | - | 1 - 2 | 641 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |