Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: per | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272265.1 | NP_001259194.1 | 6 - 7 | 3814 | 93 | |
NM_080317.2 | NP_525056.2 | 6 - 7 | 3814 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |