Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pp2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297938.1 | NP_001284867.1 | 2 - 3 | 2243 | 91 | |
NM_080335.3 | NP_525074.2 | 2 - 3 | 2243 | 91 | ||
GenBank mRNA | AY069593.1 | - | 2 - 3 | 2212 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |