Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: up | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014737.2 | NP_001014737.1 | 3 - 4 | 186 | 75 | |
NM_001169271.2 | NP_001162742.1 | 3 - 4 | 186 | 75 | ||
NM_001272594.2 | NP_001259523.1 | 3 - 4 | 183 | 75 | ||
NM_080349.3 | NP_525088.2 | 3 - 4 | 186 | 75 | ||
GenBank mRNA | BT125908.1 | - | 3 - 4 | 165 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |