Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080360.5 | NP_525099.3 | 2 - 3 | 251 | 78 | |
NM_164496.1 | NP_722830.2 | 3 - 4 | 202 | 78 | ||
GenBank mRNA | AY051789.1 | - | 2 - 3 | 235 | 78 | |
AY071846.1 | - | 2 - 3 | 200 | 78 | ||
BT053687.1 | - | 2 - 3 | 251 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |