Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdgC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370029.1 | NP_001356983.1 | 5 - 6 | 830 | 107 | |
NM_080490.4 | NP_536738.3 | 6 - 7 | 1679 | 107 | ||
NM_176366.2 | NP_788544.2 | 6 - 7 | 1317 | 107 | ||
NM_176367.3 | NP_788545.3 | 6 - 7 | 1492 | 107 | ||
NM_176368.2 | NP_788546.1 | 5 - 6 | 1007 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |