Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298460.1 | NP_001285389.1 | 3 - 4 | 1369 | 57 | |
NM_001298461.1 | NP_001285390.1 | 3 - 4 | 1373 | 57 | ||
NM_133014.3 | NP_573242.2 | 3 - 4 | 1369 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |