Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299597.1 | NP_001286526.1 | 8 - 9 | 2006 | 80 | |
NM_001299598.1 | NP_001286527.1 | 8 - 9 | 1757 | 80 | ||
NM_145335.3 | NP_663308.1 | 8 - 9 | 1695 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Drosophila melanogaster |