Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272938.2 | NP_001259867.1 | 5 - 6 | 522 | 62 | |
NM_001298631.1 | NP_001285560.1 | 5 - 6 | 608 | 62 | ||
NM_175946.2 | NP_787960.1 | 5 - 6 | 522 | 62 | ||
GenBank mRNA | AY084185.1 | - | 5 - 6 | 519 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |