Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058017.5 | NP_477365.1 | 1 - 2 | 690 | 53 | |
GenBank mRNA | BT050466.1 | - | 1 - 2 | 688 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |