Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169462.2 | NP_001162933.1 | 9 - 10 | 2105 | 135 | |
NM_058074.4 | NP_477422.2 | 10 - 11 | 1948 | 135 | ||
NM_058075.4 | NP_477423.1 | 10 - 11 | 1836 | 135 | ||
NM_134320.3 | NP_599147.1 | 10 - 11 | 1836 | 135 | ||
NM_134321.3 | NP_599148.1 | 8 - 9 | 1606 | 135 | ||
NM_134322.3 | NP_599149.1 | 8 - 9 | 1606 | 135 | ||
GenBank mRNA | AF144232.1 | - | 9 - 10 | 1379 | 135 | |
AF161259.1 | - | 8 - 9 | 1617 | 135 | ||
BT025053.1 | - | 9 - 10 | 2034 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |