Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tehao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298972.1 | NP_001285901.1 | 1 - 2 | 1179 | 84 | |
NM_058090.4 | NP_477438.1 | 1 - 2 | 1179 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |