Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: out | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298506.1 | NP_001285435.1 | 2 - 3 | 631 | 62 | |
NM_133150.3 | NP_573378.1 | 2 - 3 | 631 | 62 | ||
GenBank mRNA | AF184230.1 | - | 2 - 3 | 564 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |