Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dop2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014759.2 | NP_001014759.1 | 6 - 7 | 1574 | 104 | |
NM_001298548.1 | NP_001285477.1 | 7 - 8 | 2971 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |