Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dop2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014757.3 | NP_001014757.2 | 3 - 4 | 1958 | 75 | |
NM_001014758.3 | NP_001014758.2 | 3 - 4 | 1958 | 75 | ||
NM_001014760.3 | NP_001014760.2 | 3 - 4 | 1958 | 75 | ||
NM_001031909.2 | NP_001027080.2 | 3 - 4 | 1958 | 75 | ||
NM_001298548.1 | NP_001285477.1 | 2 - 3 | 1842 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |