Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp6v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298570.1 | NP_001285499.1 | 3 - 4 | 2356 | 65 | |
NM_001298571.1 | NP_001285500.1 | 2 - 3 | 2117 | 65 | ||
NM_134559.3 | NP_608403.1 | 2 - 3 | 2582 | 65 | ||
GenBank mRNA | BT031316.1 | - | 2 - 3 | 1754 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |