Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164382.2 | NP_722629.1 | 2 - 3 | 750 | 73 | |
GenBank mRNA | AY121696.1 | - | 2 - 3 | 746 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |