Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298636.1 | NP_001285565.1 | 1 | 540 | 59 | |
NM_164435.3 | NP_722725.1 | 1 | 540 | 59 | ||
GenBank mRNA | BT023807.1 | - | 1 | 537 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |