Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fred | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258937.2 | NP_001245866.1 | 8 - 9 | 4486 | 106 | |
NM_134968.4 | NP_608812.3 | 9 - 10 | 4858 | 106 | ||
NM_164569.4 | NP_722954.3 | 9 - 10 | 4855 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |