Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tif-IA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176070.3 | NP_788084.1 | 1 - 2 | 435 | 125 | |
GenBank mRNA | AY061123.1 | - | 1 - 2 | 389 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |