Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tif-IA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176070.3 | NP_788084.1 | 5 - 6 | 1838 | 105 | |
GenBank mRNA | AY061123.1 | - | 5 - 6 | 1792 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |