Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spn-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057392.5 | NP_476740.1 | 2 - 3 | 376 | 73 | |
GenBank mRNA | BT044133.1 | - | 2 - 3 | 363 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |