Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp99A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202380.2 | NP_001189309.1 | 5 - 6 | 1856 | 61 | |
NM_057468.3 | NP_476816.1 | 9 - 10 | 2795 | 61 | ||
NM_143434.5 | NP_651691.4 | 16 - 17 | 3812 | 61 | ||
NM_170409.3 | NP_733288.1 | 10 - 11 | 2926 | 61 | ||
NM_206579.3 | NP_996302.1 | 10 - 11 | 2971 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |