Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276214.1 | NP_001263143.1 | 2 - 3 | 1144 | 101 | |
NM_057519.4 | NP_476867.1 | 3 - 4 | 1192 | 101 | ||
GenBank mRNA | BT031258.1 | - | 3 - 4 | 656 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Drosophila melanogaster |