Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276125.1 | NP_001263054.1 | 5 - 6 | 2352 | 56 | |
NM_057545.4 | NP_476893.1 | 5 - 6 | 2352 | 56 | ||
GenBank mRNA | BT133277.1 | - | 5 - 6 | 1755 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |