Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gycalpha99B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057740.4 | NP_477088.2 | 3 - 4 | 656 | 60 | |
GenBank mRNA | BT133227.1 | - | 3 - 4 | 638 | 60 | |
U27117.1 | - | 3 - 4 | 617 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |