Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ask1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057741.4 | NP_477089.3 | 3 - 4 | 698 | 62 | |
NM_206518.2 | NP_996240.1 | 2 - 3 | 1270 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |