Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: opa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382012.1 | NP_001368991.1 | 2 - 3 | 1417 | 77 | |
NM_079504.4 | NP_524228.2 | 2 - 3 | 1417 | 77 | ||
GenBank mRNA | BT029391.1 | - | 2 - 3 | 1385 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |