Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260058.2 | NP_001246987.1 | 3 - 4 | 357 | 86 | |
NM_079554.4 | NP_524278.1 | 2 - 3 | 237 | 86 | ||
NM_169220.3 | NP_731241.1 | 2 - 3 | 245 | 86 | ||
NM_169221.3 | NP_731242.1 | 2 - 3 | 196 | 86 | ||
GenBank mRNA | AF078907.1 | - | 2 - 3 | 204 | 86 | |
AY060424.1 | - | 1 - 2 | 1556 | 86 | ||
BT003779.1 | - | 2 - 3 | 248 | 86 | ||
BT012325.1 | - | 2 - 3 | 188 | 86 | ||
BT044500.1 | - | 2 - 3 | 243 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |