Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275482.1 | NP_001262411.1 | 4 - 5 | 826 | 65 | |
NM_001275483.1 | NP_001262412.1 | 4 - 5 | 826 | 65 | ||
NM_079564.4 | NP_524288.3 | 4 - 5 | 826 | 65 | ||
NM_169274.2 | NP_731341.1 | 4 - 5 | 826 | 65 | ||
NM_169275.2 | NP_731342.1 | 2 - 3 | 267 | 65 | ||
GenBank mRNA | BT003462.1 | - | 4 - 5 | 824 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |