Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275598.2 | NP_001262527.1 | 7 - 8 | 1150 | 77 | |
NM_001275599.1 | NP_001262528.1 | 7 - 8 | 1454 | 77 | ||
NM_079616.4 | NP_524340.2 | 7 - 8 | 1093 | 77 | ||
NM_169495.4 | NP_731771.3 | 7 - 8 | 1430 | 77 | ||
GenBank mRNA | AY129457.1 | - | 7 - 8 | 1090 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |