Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275663.1 | NP_001262592.1 | 3 - 4 | 909 | 78 | |
NM_001275664.1 | NP_001262593.1 | 3 - 4 | 1227 | 78 | ||
NM_001300408.1 | NP_001287337.1 | 2 - 3 | 854 | 78 | ||
NM_079637.5 | NP_524361.4 | 3 - 4 | 937 | 78 | ||
NM_169644.2 | NP_732012.1 | 3 - 4 | 990 | 78 | ||
GenBank mRNA | BT044580.1 | - | 3 - 4 | 937 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |