Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-IIa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300446.1 | NP_001287375.1 | 5 - 6 | 1710 | 93 | |
NM_079660.4 | NP_524384.3 | 6 - 7 | 1865 | 93 | ||
GenBank mRNA | AF325713.1 | - | 6 - 7 | 1664 | 93 | |
AY118373.1 | - | 6 - 7 | 1863 | 93 | ||
BT023786.1 | - | 5 - 6 | 961 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |