Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275880.2 | NP_001262809.1 | 2 - 3 | 1388 | 84 | |
NM_001300525.1 | NP_001287454.1 | 2 - 3 | 977 | 84 | ||
NM_001300526.1 | NP_001287455.1 | 4 - 5 | 1444 | 84 | ||
NM_079713.3 | NP_524437.2 | 2 - 3 | 1768 | 84 | ||
NM_169961.3 | NP_732646.1 | 4 - 5 | 1005 | 84 | ||
NM_169962.3 | NP_732647.1 | 2 - 3 | 1569 | 84 | ||
GenBank mRNA | BT016096.1 | - | 4 - 5 | 978 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |