Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpI12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079715.3 | NP_524439.1 | 2 - 3 | 416 | 75 | |
GenBank mRNA | BT050591.1 | - | 2 - 3 | 362 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |