Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104392.3 | NP_001097862.1 | 13 - 14 | 2921 | 72 | |
NM_001104393.3 | NP_001097863.1 | 13 - 14 | 1490 | 72 | ||
NM_001170210.2 | NP_001163681.1 | 18 - 19 | 3086 | 72 | ||
NM_001275882.1 | NP_001262811.1 | 17 - 18 | 3032 | 72 | ||
NM_001382187.1 | NP_001369027.1 | 10 - 11 | 2231 | 72 | ||
NM_079717.5 | NP_524441.2 | 17 - 18 | 3032 | 72 | ||
NM_169970.3 | NP_732661.1 | 12 - 13 | 3016 | 72 | ||
NM_169971.3 | NP_732662.2 | 14 - 15 | 2975 | 72 | ||
GenBank mRNA | BT044550.1 | - | 13 - 14 | 2881 | 72 | |
BT046172.1 | - | 17 - 18 | 3016 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |