Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNApol-epsilon255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079738.3 | NP_524462.2 | 3 - 4 | 6469 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |