Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glob3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141340.3 | NP_649597.3 | 1 - 2 | 275 | 123 | |
GenBank mRNA | BT022666.1 | - | 1 - 2 | 275 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |