Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043221.4 | NP_001036686.3 | 5 - 6 | 1129 | 79 | |
NM_001300302.1 | NP_001287231.1 | 4 - 5 | 1006 | 79 | ||
NM_141518.4 | NP_649775.2 | 5 - 6 | 1175 | 79 | ||
NM_169212.3 | NP_731221.2 | 6 - 7 | 1295 | 79 | ||
GenBank mRNA | BT044586.1 | - | 3 - 4 | 932 | 79 | |
BT126358.1 | - | 4 - 5 | 990 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |