Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141884.4 | NP_650141.2 | 2 - 3 | 765 | 67 | |
GenBank mRNA | BT044493.1 | - | 2 - 3 | 833 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster Inventoried |