Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: f-cup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104302.2 | NP_001097772.1 | 6 - 7 | 1483 | 71 | |
NM_001104303.2 | NP_001097773.1 | 4 - 5 | 610 | 71 | ||
NM_001104304.2 | NP_001097774.1 | 3 - 4 | 627 | 71 | ||
NM_001275615.1 | NP_001262544.1 | 5 - 6 | 966 | 71 | ||
NM_001275616.1 | NP_001262545.1 | 3 - 4 | 878 | 71 | ||
NM_001275617.1 | NP_001262546.1 | 3 - 4 | 878 | 71 | ||
NM_001275618.1 | NP_001262547.1 | 4 - 5 | 1395 | 71 | ||
NM_142049.3 | NP_650306.1 | 3 - 4 | 878 | 71 | ||
NM_169550.2 | NP_731852.2 | 5 - 6 | 966 | 71 | ||
GenBank mRNA | AF184226.1 | - | 3 - 4 | 850 | 71 | |
BT012334.1 | - | 6 - 7 | 1432 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |